Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Graphite, powder, TECHNICAL

Graphite, powder, TECHNICAL
Supplier:  VWR Chemicals
Certificates
Graphite, powder, TECHNICAL

2 Options Available Below

Product Details & Documents
  • CAS Number:7782-42-5
  • Formula:C
  • MDL Number:MFCD00144065
  • Molecular Weight:12.01 g/mol
  • Merck Index:12,04560
  • Melting Pt:3827 °C
  • Density:2.09…2.23 g/cm³ (20 °C)
  • EINECS:231-955-3
Specifications for Product Family

Specification Test Results

  • Identification:
    Passes test
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.