Order Entry
Austria
ContactUsLinkComponent
 
Graphite ≥99.998% (metals basis), rods, Length: 30.5 cm (12 in), Ø 13cm (5.125 in)
Catalog # 43649.KF
CAS Number:  
Graphite ≥99.998% (metals basis), rods, Length: 30.5 cm (12 in), Ø 13cm (5.125 in)
Catalog # 43649.KF
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
Merck Index:12,04560
Melting Pt:3827 °C
Density:2.09…2.23 g/cm³ (20 °C)
 :12.01 g/mol
Storage Temperature:Ambient
EINECS:231-955-3
CAS Number:7782-42-5
Formula:C
MDL Number:MFCD00144065