

Graphite (natural) ≥99.9995% (metals basis), powder -200 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7782-42-5
:
Formula:C
MDL Number:MFCD00144065
Merck Index:12,04560
Melting Pt:3827 °C
Density:2.09…2.23 g/cm³ (20 °C)
EINECS:231-955-3
UN:1325
ADR:4.1,III
:12.01 g/mol
:GHS02
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together