Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Graphite
Graphite, plate, Thickness: 2.5 cm (1.0 in)
Graphite, plate, Thickness: 2.5 cm (1.0 in)
00882.VE
:
Thermo Fisher Scientific
:
Graphite, plate, Thickness: 2.5 cm (1.0 in)
00882.VE
:
Thermo Fisher Scientific
:
Pk:
1
Seq:
0001
:
:
12,04560
:
3827 °C
:
2.09…2.23 g/cm³ (20 °C)
:
12.01 g/mol
:
Ambient
:
231-955-3
:
7782-42-5
:
C
:
MFCD00144065