Order Entry
Austria
Orders LinkContactUsLinkComponent
Graphite, powder

Graphite, powder

Glass bottle, 100 g
Catalog # ACRO385031000
Graphite, powder
Catalog # ACRO385031000
CAS Number:  7782-42-5
Graphite, powder
Graphite, powder

Glass bottle, 100 g

CAS Number:  7782-42-5
Price
64,40 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:7782-42-5
  • Formula:C
  • MDL Number:MFCD00144065
  • Molecular Weight:12.01 g/mol
  • Merck Index:12,04560
  • Melting Pt:3827 °C
  • Density:2,09 to 2.23 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-955-3

Documents

Specifications

Specification Test Results

  • Appearance:
    Black Powder
  • Ash:
    ≤0.8 %
  • Carbon (C):
    ≥99.0 %
  • Sulfur (S):
    ≤0.05 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.