Magnesium ≥99.98% (trace metals basis), chips 6 - 35 mesh, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7439-95-4
:
Formula:Mg
MDL Number:MFCD00085308
:24.3 g/mol
Merck Index:13,05675
Boiling Pt:1107 °C (1013 hPa)
Melting Pt:651 °C
Density:1.74 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1869
ADR:4.1,III
:M-Clarity™ quality level = MQ100
EINECS:231-104-6
Frequently Bought Together