Order Entry
Austria
ContactUsLinkComponent
Magnesium, turnings for grignard-reaction, for synthesis, Sigma-Aldrich®
Flammable
Magnesium, turnings for grignard-reaction, for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Magnesium, turnings for grignard-reaction, for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) (slow decomposition)
  • Melting Point:
    651 °C
  • Molar Mass:
    24.31 g/mol
  • Boiling Point:
    1107 °C
  • Flash Point:
    500 °C
  • Density:
    1.75 g/cm3 (20 °C)
  • Ignition Point:
    450 °C
  • Hazard Class:
    4.1
  • Hazard Class Description:
    Flammable solid

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together