Order Entry
Austria
ContactUsLinkComponent
 
 
Magnesium ≥99.5%, foil, Width: 3 mm, Thickness: 0.15-0.30 mm, Supelco®
 :  Merck
 
 
Magnesium ≥99.5%, foil, Width: 3 mm, Thickness: 0.15-0.30 mm, Supelco®
 :  Merck

 

  • Assay (complexometric):
    ≥ 99.5 %
  • Substances insoluble in hydrochloric acid:
    ≤ 0.05 %
  • Cu (Copper):
    ≤ 0.005 %
  • Fe (Iron):
    ≤ 0.05 %
  • Pb (Lead):
    ≤ 0.01 %

 

Frequently Bought Together