Order Entry
Austria
ContactUsLinkComponent
Magnesium, cubes ∼ 19 mm (0.75 in) each side, Length: 19mm (0.75in)
Magnesium, cubes ∼ 19 mm (0.75 in) each side, Length: 19mm (0.75in)
Catalog # 45119.KF
CAS Number:  
Magnesium, cubes ∼ 19 mm (0.75 in) each side, Length: 19mm (0.75in)
Catalog # 45119.KF
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:7439-95-4
 :
Formula:Mg
MDL Number:MFCD00085308
 :24.3 g/mol
Merck Index:13,05675
Boiling Pt:1107 °C (1013 hPa)
Melting Pt:651 °C
Density:1.74 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-104-6
UN:1869
ADR:4.1,III