Order Entry
Austria
ContactUsLinkComponent
Magnesium ≥99.95% (metals basis), slug, unannealed, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Magnesium ≥99.95% (metals basis), slug, unannealed, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Catalog # 43296.09
CAS Number:  
Magnesium ≥99.95% (metals basis), slug, unannealed, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Catalog # 43296.09
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :24.3 g/mol
Merck Index:13,05675
Boiling Pt:1107 °C (1013 hPa)
Melting Pt:651 °C
Density:1.74 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-104-6
UN:1869
ADR:4.1,III
CAS Number:7439-95-4
Formula:Mg
MDL Number:MFCD00085308