Water-¹⁸O, 10 atom% ¹⁸O, Sigma-Aldrich®
: Merck
: Water-18O, 18O-Water
:14314-42-2
:
:H₂¹⁸O
:MFCD00064503
:100 °C (lit.)
:0 °C (lit.)
:1.11 g/ml (20 °C) (lit.)
:20.02 g/mol
:M-Clarity™ quality level = MQ200
- Environmentally Preferable:
- Proton NMR Spectrum:Conforms to Structure
- Isotopic Assay (CA):10 atom% per 18-O (typically 10.0 - 10.5)
Frequently Bought Together