Order Entry
Austria
ContactUsLinkComponent
RNA isolation, NucleoSpin® RNA Midi Kit
RNA isolation, NucleoSpin® RNA Midi Kit
Catalog # MANA740962.20
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
RNA isolation, NucleoSpin® RNA Midi Kit
Catalog # MANA740962.20
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    NucleoSpin® RNA Midi, 20 preparations
  • Yield:
    180 µg (10⁷ HeLa cells); 620 µg (4 x 10⁷ HeLa cells)

Specifications

About this item

NucleoSpin® RNA Midi spin kit is used for the isolation of RNA of highest integrity.

  • Up to 600 μg ready to use RNA
  • Used for RNase protection assays, RNA isolation and separations
  • Sample material: < 5×107 cultured cells, < 1010 bacterial cells, < 3×108 yeast cells, <200 mg human/animal tissue

The rDNase included for on-column digestion results in easy and efficient removal of genomic DNA. The NucleoSpin® filters midi (shredders) gives an efficient sample homogenisation and reduction of viscosity.

Parameters
Binding capacity: 700 µg
Elution volume: 500 – 1000 µl
Format: Midi spin columns
Fragment size: >200 nt
Sample type: Bacterial cells;Cultured cells;Yeast cultures;Human and animal fatty tissues
Time: 80 min/4 preps
Yield: 180 µg (10⁷ HeLa cells); 620 µg (4 x 10⁷ HeLa cells)

Delivery: Kit includes NucleoSpin® RNA midi columns with collection tubes (15 ml), collection tubes (15 ml), NucleoSpin® filters midi, buffers, RNase-free rDNase.