Order Entry
Austria
ContactUsLinkComponent
Reference standard for food and environment analysis, disperse yellow 49, Dr Ehrenstorfer
Reference standard for food and environment analysis, disperse yellow 49, Dr Ehrenstorfer
Catalog # EHERC12972349
Supplier:  EHRENSTORFER
CAS Number:  
Reference standard for food and environment analysis, disperse yellow 49, Dr Ehrenstorfer
Catalog # EHERC12972349
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    0,1 g
  • CAS No.:
    6858-49-7
  • Description:
    Disperse yellow 49, 100 mg
  • Formula:
    C₂₂H₂₂N₄O₂
  • Description:
    Disperse yellow 49

Specifications

About this item