Order Entry
Austria
ContactUsLinkComponent
Guide roll, lockable
Guide roll, lockable
  BOCH100
 :  BOCHEM
undefined
Guide roll, lockable
  BOCH100
 :  BOCHEM

 

  • Description:
    Guide roll, lockable, 26,9 mm, 70000 g
  • Pk:
    1

 

 

Guide roll are made of 18/10 stainless steel.

  • Diameter: 100 mm
  • Height: 121 mm