ContactUsLinkComponent
Wound retractor, Irwin, Aesculap®
Catalog # AESCBV247R
Supplier:  Aesculap
Wound retractor, Irwin, Aesculap®
Catalog # AESCBV247R
Supplier:  Aesculap
Specifications
  • Description:
    Abdominal retractor, sharp, with 4×4 teeth
  • Length:
    175 mm
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Self-retaining, abdominal retractor, type Irwin.
  • Sharp
  • 4×4 teeth