Orders LinkContactUsLinkComponent
Conductivity cells, IDS
Conductivity cells, IDS
Supplier:   WTW
Conductivity cells, IDS
Supplier:  WTW
Certificates
Conductivity cells, IDS
Conductivity cells, IDS
8 Options Available Below
Product Details & Documents
High quality, enhanced sensor technology combined with most up to date measurement electronics. For use with IDS meters.
  • Intelligent: Sensors are automatically recognised by the meter, each sensor is individually identified
  • Digital: Measurement signals are processed digitally, providing very accurate data
  • IDS cells store their serial number and calibration records
  • TetraCon® 925: 4-electrode measuring cell ideal for medium and high conductivity applications
  • LR 925/01: Ideal for pure water and low conductivity applications
  • TetraCon® 925/LV: Graphite 4-electrode cell for small volumes and viscous samples
The electrodes can provide the following data, shown on the screen of the digital meter:
- Model name
- Serial number
- Calibration status (+++, ++, +, -, blank)
- The calibration history includes the last 10 calibrations, including buffers used, mV values, etc.
Ordering information: Various cable lengths available.
Product Family Options
Product Information
  • Type
    Conductivity
    Cell constant
    Temp.
    Material
    Ø×L
    Connection
    Availability
    Price
  • 663-0152
    IDS conductivity cell, TetraCon® 925, 4-cell graphite
    IDS conductivity cell, TetraCon® 925, 4-cell graphite
    TetraCon® 925, 4-cell graphite
    10 µS/cm - 2000 mS/cm
    0,475 cm-1
    0...+100 °C
    Epoxy
    15,3×120 mm
    IDS-plug, 1,5 m cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 639,00 €
    639,00 €
    1 * 1 ST
       639,00 €
  • 663-0153
    IDS conductivity cell, TetraCon® 925-3, 4-cell graphite
    IDS conductivity cell, TetraCon® 925-3, 4-cell graphite
    TetraCon® 925-3, 4-cell graphite
    10 µS/cm - 2000 mS/cm
    0,475 cm-1
    0...+100 °C
    Epoxy
    15,3×120 mm
    IDS-plug, 3 m cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 657,00 €
    657,00 €
    1 * 1 ST
       657,00 €
  • 665-0304
    IDS conductivity cell, TetraCon® 925-P, 4-cell graphite, wireless ready
    IDS conductivity cell, TetraCon® 925-P, 4-cell graphite, wireless ready
    TetraCon® 925-P, 4-cell graphite, wireless ready
    10 µS/cm - 2000 mS/cm
    0,475 cm-1
    0...+100 °C
    Epoxy
    15,3×120 mm
    Plug head for AS/IDS-x cable or wireless module
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 660,00 €
    660,00 €
    1 * 1 ST
       660,00 €
  • 665-0408
    IDS conductivity cell, TetraCon® 925/LV, 4-cell, fork design, for low volumes + viscous samples
    IDS conductivity cell, TetraCon® 925/LV, 4-cell, fork design, for low volumes + viscous samples
    TetraCon® 925/LV, 4-cell, fork design (for viscous samples)
    1 µS/cm - 2000 mS/cm
    0,469 cm-1
    0...+100 °C
    Epoxy
    15,3×120 mm
    IDS-plug, 1,5 m cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 863,00 €
    863,00 €
    1 * 1 ST
       863,00 €
  • 665-0474
    IDS conductivity cell, TetraCon® 925/LV-P, 4-cell, fork design, for low volumes + viscous samples, wireless ready
    IDS conductivity cell, TetraCon® 925/LV-P, 4-cell, fork design, for low volumes + viscous samples, wireless ready
    TetraCon® 925/LV-P, 4-cell, fork design (for viscous samples), wireless ready
    1 µS/cm - 2000 mS/cm
    0,469 cm-1
    0...+100 °C
    Epoxy
    15,3×120 mm
    Plug head for AS/IDS-x cable or wireless module
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 886,00 €
    886,00 €
    1 * 1 ST
       886,00 €
  • 663-0156
    IDS conductivity cell, LR 925/01, 2-cell stainless steel, incl. flow container
    IDS conductivity cell, LR 925/01, 2-cell stainless steel, incl. flow container
    LR 925/01, 2-cell stainless steel, incl. flow container
    0,01 - 200 µS/cm
    0,100 cm-1
    0...+100 °C
    Stainless steel
    12×120 mm
    IDS plug, 1,5 m cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 884,00 €
    884,00 €
    1 * 1 ST
       884,00 €
  • 665-0475
    IDS conductivity cell, TetraCon® 925/C, 4-cell graphite, PEEK head
    IDS conductivity cell, TetraCon® 925/C, 4-cell graphite, PEEK head
    TetraCon® 925/C, 4-cell graphite, PEEK head
    1 µS/cm - 2000 mS/cm
    0,475 cm-1
    0...+100 °C
    PEEK
    15,3×120 mm
    IDS-plug, 1,5 m cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 991,00 €
    991,00 €
    1 * 1 ST
       991,00 €
  • WTWA301722
    IDS conductivity cell, LR 925/01-P, 2-cell stainless steel, incl. flow container
    IDS conductivity cell, LR 925/01-P, 2-cell stainless steel, incl. flow container
    LR 925/01-P, 4-cell graphite, PEEK head
    0,01 - 200 µS/cm
    0,100 cm⁻¹
    0...+100 °C
    Stainless steel
    12×120 mm
    Plug head for AS/IDS-x cable or IDS WLM-S wireless module
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 976,00 €
    976,00 €
    1 * 1 ST
       976,00 €
Connection cables for IDS sensors
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • AS/IDS-1,5, connection cable for IDS sensors with waterproof plug head
    1,5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 114,00 €
    114,00 €
    1 * 1 ST
       114,00 €
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • AS/IDS-10, connection cable for IDS sensors with waterproof plug head
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 196,00 €
    196,00 €
    1 * 1 ST
       196,00 €
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • AS/IDS-100, connection cable for IDS sensors with waterproof plug head
    100 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 100,00 €
    1 100,00 €
    1 * 1 ST
       1 100,00 €
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • AS/IDS-60, connection cable for IDS sensors with waterproof plug head
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 846,00 €
    846,00 €
    1 * 1 ST
       846,00 €
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • AS/IDS-20, connection cable for IDS sensors with waterproof plug head
    20 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 245,00 €
    245,00 €
    1 * 1 ST
       245,00 €
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • AS/IDS-40, connection cable for IDS sensors with waterproof plug head
    40 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 703,00 €
    703,00 €
    1 * 1 ST
       703,00 €
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • AS/IDS-15, connection cable for IDS sensors with waterproof plug head
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 224,00 €
    224,00 €
    1 * 1 ST
       224,00 €
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • AS/IDS-3, connection cable for IDS sensors with waterproof plug head
    3 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 155,00 €
    155,00 €
    1 * 1 ST
       155,00 €