pH indicator, bromo cresol purple tablets, Lovibond®
Supplier: Lovibond
Certificates
About this item
Indicator for pH range 5,2 to 6,8.
Can be used for measuring pH in conjunction with the Comparator 2000+ system.
Frequently Bought Together
Indicator for pH range 5,2 to 6,8.
Can be used for measuring pH in conjunction with the Comparator 2000+ system.