Autophagy library, Screen-Well®
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Structurally and mechanistically different compounds for studying the roles of pro- and anti-autophagic molecules in cells and in vitro applications.
The Screen-well® autophagy library contains 94 compounds with defined autophagy-inducing or -inhibitory activity. Compounds are dissolved in DMSO at 10 mM or 1 mM and aliquoted into deep-well plates at 100 or 500 µl per well. A variety of structurally and mechanistically different compound classes are included. A useful tool for studying the roles of pro- and anti-autophagic molecules in cells as well as for use in in vitro applications.
Ordering information: Contains 94 compounds with defined autophagy-inducing or -inhibitory activity. A variety of structurally and mechanistically different compound classes are included.
Specifications
- Concentration:DMSO solutions (10 mM ) except for Bafilomycin A which is provided at 1 mM in DMSO.