Serotonergic ligand library, Screen-Well®
Supplier: ENZO LIFE SCIENCES
New Product
Certificates
About this item
The Screen-Well® serotonergic ligands plate contains 79 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The serotonergic ligands plate is 1 plate of the larger neurotransmitter library which contains over 680 ligands.
Specifications
- Source:
- Species:
- Environmentally Preferable:
Specifications
Frequently Bought Together