Ion channel ligand library, Screen-Well®
The Screen-Well® ion channel ligand library contains 71 ion channel blockers and openers for use in characterising and identifying ion channels in individual cells or tissue.
- Concentration:10 mM in DMSO
Frequently Bought Together