Order Entry
Austria
ContactUsLinkComponent
STT negative control for gelatinase
STT negative control for gelatinase
Catalog # ENZOBMLPI1370005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
STT negative control for gelatinase
Catalog # ENZOBMLPI1370005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    03
  • Protein/peptide name:
    STT Negative Control for Gelatinase
  • Purity:
    >95% (HPLC)
  • Tested applications:
    Negative control

Specifications

About this item

MMP inhibitor

STTHWGFTLS is ten times less potent than, thus a negative control for CTTHWGFTLC (Prod. No. BML-PI136), which is a specific inhibitor of the gelatinases MMP-2 and MMP-9. Can be used in in vitro studies, zymography and in situ zymography, in tissue culture, and in vivo.