PPACK
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Potent irreversible thrombin inhibitor. Inhibits thrombin activation of progelatinase A. Pretreatment with PPACK inhibits thrombin-induced cerebral thromboembolism in vivo.
- Thrombin inhibitor
White to off-white solid.
Soluble in water.
Specifications
- Protein synonyms:D-Phe-Pro-Arg-CMK
- Protein/peptide name:PPACK