Order Entry
Austria
ContactUsLinkComponent
RGD peptide
RGD peptide
Catalog # ENZOBMLP7000005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
RGD peptide
Catalog # ENZOBMLP7000005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    02
  • Protein/peptide name:
    RGD Peptide
  • Purity:
    >97% (HPLC)

Specifications

About this item

Integrin ligand

Induces apoptosis in various cell types and cell lines at 1mM by a mechanism involving cellular uptake and possible direct stimulation of procaspase-3 autoprocessing. Does not induce apoptosis in the breast cancer line MCF-7, in which there is a functional deletion of the procaspase-3 gene. Contains the motif RGD (Arg-Gly-Asp) which is found in many integrin ligands. Inhibits cell adhesion of COS-7 cells to fibronectin 80-90% at 1 mM.