pp60c-src C-terminal phosphoregulatory peptide
Catalog # ENZOBMLP3010001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Seq:02
- Protein/peptide name:pp60c-src C-terminal phosphoregulatory Peptide
- Purity:>98%
Specifications
About this item
Src kinase inhibitor
This phosphopeptideis the C-terminal negative regulatory domain of c-Src (521-533) which includes the negative regulatory site (Tyr527). This peptide, when tyrosine-phosphorylated, binds to the internal SH2 domain.