Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Fluor-de-Lys® Developer II concentrate

Fluor-de-Lys® Developer II concentrate, 1,25 mL
Catalog # ENZOBMLKI1761250
Supplier:   Enzo Life Sciences
Fluor-de-Lys® Developer II concentrate
Catalog # ENZOBMLKI1761250
Supplier:  Enzo Life Sciences
CAS Number:  
Fluor-de-Lys® Developer II concentrate

Fluor-de-Lys® Developer II concentrate, 1,25 mL

CAS Number:  
Price
261,00 €
Log in for availability
1 * 1,25 mL
Product Details & Documents

FLUOR DE LYS® Developer II is an essential reagent for signal generation in assays done with the FLUOR DE LYS®-HDAC8, FLUOR DE LYS®-SIRT1 and FLUOR DE LYS®-SIRT2 substrates. Deacetylation of the FLUOR DE LYS® substrate sensitises it so that, in a second step, treatment with the FLUOR DE LYS® Developer II produces a fluorophore. Typically, an ‘inhibitor stop’ will be included in the 1X Developer dilution (e.g. nicotinamide for sirtuins).

Ordering information: Quantity: NOTE: This product is supplied packaged in 5 tubes (250 µl per tube) for a total volume of 1,25 ml. This packaging is used for optimal stability of the developer.

Specifications

Specifications

  • Pk:
    1,25 mL
  • Description:
    Fluor-de-Lys® Developer II concentrate
  • Cat. No.:
    ENZOBMLKI1761250
  • Concentration:
    5X concentrate (1,25 ml). After dilution (1X), sufficient for 100 assays (50 µl reaction volume)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.