Order Entry
Austria
Orders LinkContactUsLinkComponent
Fluor-de-Lys® Developer I concentrate

Fluor-de-Lys® Developer I concentrate

Fluor-de-Lys® Developer I concentrate, 300 µl
Catalog # ENZOBMLKI1050300
Supplier:   Enzo Life Sciences
Fluor-de-Lys® Developer I concentrate
Catalog # ENZOBMLKI1050300
Supplier:  Enzo Life Sciences
CAS Number:  
Fluor-de-Lys® Developer I concentrate

Fluor-de-Lys® Developer I concentrate, 300 µl

CAS Number:  
Price
224,00 €
Log in for availability
1 * 300 µl
Product Details & Documents

FLUOR DE LYS® Developer is an essential reagent for signal generation in HDAC or sirtuin assays done with the FLUOR DE LYS® substrate. An appropriate FLUOR DE LYS® family substrate, comprising an acetylated lysine side chain, is first incubated with a sample containing HDAC activity (nuclear or cellular extract, purified enzyme, bead-bound immunocomplex, etc.). Deacetylation of the substrate sensitizes it so that, in a second step, treatment with the FLUOR DE LYS® Developer produces a fluorophore. Typically, an ‘inhibitor stop’ will be included in the 1x Developer dilution (e.g. Trichostatin A) for class I and II HDACs, nicotinamide for sirtuins).

  • An essential reagent for signal generation in HDAC or sirtuin assays performed with the FLUOR DE LYS® substrate.
Specifications

Specifications

  • Pk:
    300 µl
  • Description:
    Fluor-de-Lys® Developer I concentrate
  • Cat. No.:
    ENZOBMLKI1050300
  • Concentration:
    20X concentrate. After dilution (1X), sufficient for 100 assays (50 µl reaction volume)

Specifications