Order Entry
Austria
ContactUsLinkComponent
ARF inhibitory peptide
ARF inhibitory peptide
Catalog # ENZOBMLG4450001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
ARF inhibitory peptide
Catalog # ENZOBMLG4450001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    02
  • Protein/peptide name:
    ARF inhibitory Peptide
  • Purity:
    97%

Specifications

About this item

Ultra-pure

Sequence is ADP-ribosylation factor (ARF) mARF1p (2-17) N-terminal peptide. Peptide inhibits ARF-dependent ADP-ribosylation by cholera toxin, endoplasmic reticulum to Golgi transport, intra-Golgi transport and endosome-endosome fusion in vitro at IC50=10-40 µM.