Order Entry
Austria
ContactUsLinkComponent
(D-Pro1,2)-Retro-(D-HIV-Tat(48-57))
(D-Pro1,2)-Retro-(D-HIV-Tat(48-57))
Catalog # ENZOBMLEI3840001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
(D-Pro1,2)-Retro-(D-HIV-Tat(48-57))
Catalog # ENZOBMLEI3840001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    02
  • Protein/peptide name:
    (D-Pro1,2)-Retro-(D-HIV-Tat(48-57))
  • Purity:
    >98%

Specifications

About this item

Jnk inhibitor

Useful as a control peptide in studies using D-JNKi1 (BML-EI355). This peptide is the cell transporter, and does not contain the JIP (JNK inhibitor peptide) sequence.