Mu-Phe-hPhe-FMK
ENZOBMLEI3230001
:
- Pk:1 mg
- Seq:02
- Protein/peptide name:Mu-Phe-hPhe-FMK
- Purity:98%
Cathepsin inhibitor
Irreversible, non-toxic, cell-permeable inhibitor of cysteine proteases calpain, cathepsins B and L, and Trypanosome trypanopain. Can be used in vitro (typically 20 µM) or in vivo. Inhibits tumor formation and growth.