Cyclo [Arg-Gly-Asp-D-Phe-Val]
Catalog # ENZOBMLAM1000001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- CAS No.:137813-35-5
- Seq:02
- Storage conditions:Freezer
- Protein/peptide name:Cyclo [Arg-Gly-Asp-D-Phe-Val]
Specifications
About this item
Integrin αvβ3 antagonist. Inhibits cell adhesion events mediated by αvβ3. Inhibits angiogenesis. Induces rapid regression of histologically distinct human tumors. Induces apoptosis of angiogenic blood vessels.
- Integrin inhibitor
Lyophilized solid.
Soluble in DMSO (5 mg/ml) and PBS (1 mg/350 μl).