Caspase inhibitor set
Catalog # ENZOALX850236KI01
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 SET
- Seq:02
- Protein/peptide name:Caspase inhibitor set
- Formulation:Liquid. In DMSO.
Specifications
About this item
Synthetic peptides that irreversibly inhibit the activity of each of the caspase-family proteases. YVAD is the recognition sequence for caspase-1/ICE, DEVD is the recognition sequence for caspase-3/CPP32, VEID is the recognition sequence for caspase-6/Mch2, IETD is the recognition sequence for caspase-8/ FLICE, and VAD is the recognition sequence for a caspase(s) that functions upstream of most other caspases.
Ordering information: Contents: Contains 25 µl (2mM) each of the following inhibitors:
- Caspase-1 Inhibitor, Z-YVAD-FMK, yellow cap
- Caspase-3 Inhibitor, Z-DEVD-FMK, red cap
- Caspase-6 Inhibitor, Z-VEID-FMK, violet cap
- Caspase-8 Inhibitor, Z-IETD-FMK, green cap
- Caspase-Family Inhibitor, Z-VAD-FMK, amber cap
- Caspase-1 Inhibitor, Z-YVAD-FMK, yellow cap
- Caspase-3 Inhibitor, Z-DEVD-FMK, red cap
- Caspase-6 Inhibitor, Z-VEID-FMK, violet cap
- Caspase-8 Inhibitor, Z-IETD-FMK, green cap
- Caspase-Family Inhibitor, Z-VAD-FMK, amber cap