ContactUsLinkComponent
Ac-AEVD-CHO
Ac-AEVD-CHO
Catalog # ENZOALX260158M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Ac-AEVD-CHO
Catalog # ENZOALX260158M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Pk:
    5 mg
  • Seq:
    02
  • Protein/peptide name:
    Ac-AEVD-CHO
Specifications
About this item
Inhibitor of caspase-6, -8 and -10
Inhibitor of caspase-10. Also inhibits caspase-6 and -8.