Z-IETD-Fmk
Supplier: Enzo Life Sciences
Certificates
About this item
Caspase and granzyme inhibitor
Irreversible, cell permeable inhibitor of caspase-8 and granzyme B. Inhibits also caspase-10. The inhibitor is designed as a methyl ester to facilitate cell permeability.
Specifications
- Protein/peptide name:Z-IETD-FMK
- Purity:>99% (HPLC)
- Formulation:Liquid. In DMSO.
Specifications
Frequently Bought Together