Z-DEVD-FMK
Supplier: Enzo Life Sciences
Certificates
About this item
Inhibitor of caspase-3 and -7
Irreversible, cell permeable inhibitor of caspase-3 and -7.
Specifications
- Protein/peptide name:Z-DEVD-FMK
- Purity:>98% (HPLC)
- Formulation:__
Specifications
Frequently Bought Together