N-(1-Naphthalenylsulfonyl)-Ile-Trp-aldehyde
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Cathepsin-L inhibitor
Potent, selective and reversible inhibitor of human cathepsin L (IC50=1,9 nM). Inhibits release of Ca²⁺ and hydroxyproline from bone in vitro.
Specifications
- Protein/peptide name:N-(1-Naphthalenylsulfonyl)-Ile-Trp-aldehyde
- Purity:>97% (HPLC)