Z-LE(OMe)HD(OMe)-Fmk
Catalog # ENZOALX260076M001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Seq:01
- Protein/peptide name:Z-LE(OMe)HD(OMe)-FMK
- Purity:>95% (HPLC)
- Formulation:TFA salt.
Specifications
About this item
Caspase-9 inhibitor
Irreversible, cell permeable inhibitor of caspase-9. The carboxyl groups are OMe ester form to enhance cell permeability. If purified enzyme is used, esterase should be added for the hydrolysis of the ester groups.