Ac-VEID-CHO
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Inhibitor of caspase-6, -8 and -10
Reversible aldehyde inhibitor of caspase-6. Also inhibits caspase-8 and -10. Sequence is based on lamin A site of cleavage.
Specifications
- Protein/peptide name:Ac-VEID-CHO
- Purity:>95% (HPLC)
- Formulation:Lyophilized
Specifications
Frequently Bought Together