Order Entry
Austria
ContactUsLinkComponent
Ac-VDVAD-AMC
Ac-VDVAD-AMC
Catalog # ENZOALX260060M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Ac-VDVAD-AMC
Catalog # ENZOALX260060M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    03
  • Protein/peptide name:
    Ac-VDVAD-AMC
  • Purity:
    >98% (HPLC)

Specifications

About this item

Caspase-2 inhibitor

Fluorogenic substrate for caspase-2. Preferred cleavage site for caspase-2 (Km=80 μM).