Ac-VDVAD-CHO
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Caspase-2 inhibitor
Potent, reversible aldehyde inhibitor of caspase-2 (Ki=3,5 nM). Preferred cleavage sequence for caspase-2.
Specifications
- Protein/peptide name:Ac-VDVAD-CHO
- Purity:>94% (HPLC)
- Formulation:__
Specifications
Frequently Bought Together