Ac-DEVD-pNA
Catalog # ENZOALX260033M005
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:5 mg
- Seq:04
- Protein/peptide name:Ac-DEVD-pNA
- Purity:>97% (HPLC)
- Formulation:__
Specifications
About this item
Caspase-3 inhibitor
Colorimetric substrate for caspase-3 (CPP32) (Km=9,7 µM) and related cysteine proteases. Sequence is based on PARP (poly(ADPribose) polymerase) cleavage site Asp216 for caspase-3. Cleavage is monitored colorimetrically at 405 nm.