Ac-YVAD-AMC
Supplier: Enzo Life Sciences
Certificates
About this item
Caspase-1 inhibitor
Fluorgogenic caspase-1 substrate with a Km=14 µM. Ex.:340-360 nm, Em.:440-460 nm.Sequence is based on precursor interleukin-1beta Asp116 cleavage site for caspase-1.
Specifications
- Protein/peptide name:Ac-YVAD-AMC
- Purity:>98% (HPLC)