Order Entry
Austria
ContactUsLinkComponent
Autocamtide-2-related inhibitory Peptide
Autocamtide-2-related inhibitory Peptide
Catalog # ENZOALX151029M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Autocamtide-2-related inhibitory Peptide
Catalog # ENZOALX151029M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    04
  • Protein/peptide name:
    Autocamtide-2-related inhibitory Peptide
  • Purity:

Specifications

About this item

CaM kinase II inhibitor

Highly specific and potent inhibitor of Ca²⁺/calmodulin-dependent protein kinase II without affecting cAMP-dependent protein kinase (PKA), protein kinase C (PKC) and Ca²⁺/calmodulin-dependent protein kinase IV.