Order Entry
Austria
ContactUsLinkComponent
Human recombinant Isopeptidase T (short form) (from E. coli)
Human recombinant Isopeptidase T (short form) (from E. coli)
Catalog # ENZOBMLUW96950025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant Isopeptidase T (short form) (from E. coli)
Catalog # ENZOBMLUW96950025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    25 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Enzyme name:
    Isopeptidase T (short form)

Specifications

About this item

Produced in E. coli.