Order Entry
Austria
ContactUsLinkComponent
Human recombinant Caspase-5 (from E. coli)
Human recombinant Caspase-5 (from E. coli)
Catalog # ENZOBMLSE1715000
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant Caspase-5 (from E. coli)
Catalog # ENZOBMLSE1715000
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 Vial
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Enzyme name:
    Caspase-5
  • Enzyme activity:
    5000 U

Specifications

About this item

Produced in E. coli.