Order Entry
Austria
ContactUsLinkComponent
Apis mellifera Phospholipase A2
Apis mellifera Phospholipase A2
Catalog # ENZOBMLSE1040001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Apis mellifera Phospholipase A2
Catalog # ENZOBMLSE1040001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Species:
    Apis mellifera
  • Seq:
    02
  • Form:
    Lyophilised
  • Enzyme name:
    Phospholipase A2

Specifications

About this item

Isolated from Apis mellifera.