Order Entry
Austria
ContactUsLinkComponent
Colistin sulphate (from Bacillus colistinus)
Colistin sulphate (from Bacillus colistinus)
Catalog # ENZOALX380272M500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Colistin sulphate (from Bacillus colistinus)
Catalog # ENZOALX380272M500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    500 mg
  • CAS No.:
    1066-17-7
    1264-72-8
  • Source:
    Bacillus colistinus
  • Seq:
    02
  • Storage conditions:
    Freezer
  • Form:
    White to off-white powder
  • MDL No.:
    MFCD00146495
  • Protein synonyms:
    Polymyxin E sulfate salt
  • Protein/peptide name:
    Colistin sulphate
  • Solubility:
    Soluble in water (50 mg/ml)
  • Molecular formula:
    C₁₀₄H₂₀₆N₃₂O₄₆S₅

Specifications

About this item

Polymyxin antibiotic. Binds to lipids on the cell cytoplasmic membrane. Disrupts cell wall integrity. Effective against Gram-negative bacteria.