Order Entry
Austria
ContactUsLinkComponent
Human recombinant PD-1
Human recombinant PD-1
Catalog # ENZOENZPRT1900050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant PD-1
Catalog # ENZOENZPRT1900050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Biological activity:
    Binds to PD-L1.
  • Protein synonyms:
    CD28|PD-L1|autoimmune|tumor|apoptosis|PD-L2|infection|immune regulation|CD279|cancer|Programmed Cell Death-1|HIV|T cell
  • Protein/peptide name:
    PD-1
  • Purity:
    >90% (SDS-PAGE)
  • Molecular weight:
    ~20,5 kDa (actual is a doublet at 40 kDa and 50 kDa that is immunoactive with PD-1)
  • Formulation:
    Lyophilized

Specifications

About this item

Produced in HEK cells. Extracellular domain of PD-1 (aa 25-167) containing a 5’-His-tag, V5 epitope tag spacer, and a FLAG-tag.