Rat recombinant NGAL
ENZOENZPRT1151000
:
- Pk:1 mL
- Protein/peptide type:Recombinant
- Species:Rat
- Seq:02
- Protein synonyms:Neutrophil gelatinase-associated lipocalin|Siderocalin|Lipocalin 2|Oncogene 24p3
- Protein/peptide name:NGAL
- Concentration:400 pg/ml
- Formulation:Liquid, In 0,01 M phosphate buffer, pH 7,4, containing 0,14 M NaCl, 0,5% BSA, 0,5% Tween 20, 0,0016% Phenol red, and 0,1% Proclin 950
Produced in E. coli.