Order Entry
Austria
ContactUsLinkComponent
Corynebacterium diphtheriae Diphtheria toxin (Corynebacterium diphtheriae alpha)
Corynebacterium diphtheriae Diphtheria toxin (Corynebacterium diphtheriae alpha)
Catalog # ENZOBMLG1350001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Corynebacterium diphtheriae Diphtheria toxin (Corynebacterium diphtheriae alpha)
Catalog # ENZOBMLG1350001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Species:
    Corynebacterium diphtheriae
  • Seq:
    02
  • Protein/peptide name:
    Diphtheria toxin (Corynebacterium diphtheriae alpha)
  • Concentration:
    1 µg/µl
  • Formulation:
    Lyophilised

Specifications

About this item

From Corynebacterium diphtheriae.