Human recombinant Angiopoietin-1
Catalog # ENZOALX2018500020
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Protein synonyms:ANGPT1|Ang-1
- Protein/peptide name:Angiopoietin-1
- Purity:>85% (SDS-PAGE)
- Molecular weight:~55,6kDa
- Concentration:~1 mg/ml
- Formulation:Liquid. Sterile filtered, in 20 mM TRIS-HCl buffer, pH 8.0, 1M urea containing 5% glycerol.
Specifications
About this item
Produced in E. coli. Human Ang-1 (angiopoietin-1) is a single, non-glycosylated polypeptide chain containing 480 amino acids (20 to 498)